missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate, Monohydrate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L1125125GM

 View more versions of this product

Catalog No. 18611345


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10102-25-7
125 g
ACS
Poly Bottle
1
99%
13.0-15.0%

Chemical Identifiers

10102-25-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.