missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium sulfate monohydrate, 99%
Description
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.
Solubility
Soluble in water. 340 g/L (20 C)
Notes
Store under dry inert gas. Protect from humidity and water. Hygroscopic. Incompatible with strong oxidizing agents.
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.
Solubility
Soluble in water. 340 g/L (20 C)
Notes
Store under dry inert gas. Protect from humidity and water. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 130°C (in -H2O) |
| Quantity | 500 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.06 |
| Merck Index | 14,5541 |
| Solubility Information | Soluble in water. 340g/L (20 C) |
| Formula Weight | 127.96 (109.94 Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | Lithium sulfate monohydrate |
Safety and Handling
| Hazard Category | H302-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c |
| EINECSNumber | 233-820-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only