Learn More
Lithium sulfate monohydrate, 99+%, ACS reagent
CAS: 10102-25-7 | H2Li2O5S | 127.95 g/mol
$492.94 - $492.94
Chemical Identifiers
| CAS | 10102-25-7 |
|---|---|
| Molecular Formula | H2Li2O5S |
| Molecular Weight (g/mol) | 127.95 |
| MDL Number | MFCD00149766 |
| InChI Key | RKGLUDFWIKNKMX-UHFFFAOYSA-L |
| Synonym | lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n |
| PubChem CID | 165815 |
| IUPAC Name | dilithium;sulfate;hydrate |
| SMILES | [Li+].[Li+].O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC193395000
|
Thermo Scientific Chemicals
193395000 |
500 g | Plastic bottle |
Each for $492.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10102-25-7 | |
| 127.95 | |
| RKGLUDFWIKNKMX-UHFFFAOYSA-L | |
| 165815 | |
| [Li+].[Li+].O.[O-]S([O-])(=O)=O |
| H2Li2O5S | |
| MFCD00149766 | |
| lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n | |
| dilithium;sulfate;hydrate |
Specifications
| 860.0°C | |
| 500 g | |
| H2Li2O5S | |
| MFCD00149766 | |
| lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n | |
| RKGLUDFWIKNKMX-UHFFFAOYSA-L | |
| dilithium;sulfate;hydrate | |
| 165815 | |
| ≥99% | |
| Plastic bottle |
| 10102-25-7 | |
| 99+% | |
| Li2SO4·H2O | |
| 15,5597 | |
| Solubility in water: 295.5g/L (20°C) | |
| [Li+].[Li+].O.[O-]S([O-])(=O)=O | |
| 127.95 | |
| 127.96 | |
| ACS Reagent | |
| Lithium sulfate monohydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning
RUO – Research Use Only