Learn More
Lithium salicylate, tech.
CAS: 552-38-5 | C7H5LiO3 | 144.054 g/mol
Supplier: Thermo Scientific Chemicals 03935622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lithium salicylate | |
| (decomposition) | |
| C7H5LiO3 | |
| 100 g | |
| Soluble in water | |
| [Li+].C1=CC=C(C(=C1)C(=O)[O-])O | |
| 144.054 | |
| 144.05 | |
| Powder |
| 552-38-5 | |
| Sweet | |
| MFCD00045813 | |
| lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt | |
| PSBOOKLOXQFNPZ-UHFFFAOYSA-M | |
| lithium;2-hydroxybenzoate | |
| 23663621 | |
| Technical |
Chemical Identifiers
| 552-38-5 | |
| 144.054 | |
| PSBOOKLOXQFNPZ-UHFFFAOYSA-M | |
| 23663621 | |
| [Li+].C1=CC=C(C(=C1)C(=O)[O-])O |
| C7H5LiO3 | |
| MFCD00045813 | |
| lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt | |
| lithium;2-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
EINECSNumber : 209-011-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only