missing translation for 'onlineSavingsMsg'
Learn More

Lithium salicylate, tech.

Catalog No. AA3935636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3935636 500 g
AA3935614 25 g
AA3935622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3935636 Supplier Thermo Scientific Chemicals Supplier No. 03935636

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 552-38-5 | C7H5LiO3 | 144.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-38-5
Molecular Formula C7H5LiO3
Molecular Weight (g/mol) 144.054
MDL Number MFCD00045813
InChI Key PSBOOKLOXQFNPZ-UHFFFAOYSA-M
Synonym lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt
PubChem CID 23663621
IUPAC Name lithium;2-hydroxybenzoate
SMILES [Li+].C1=CC=C(C(=C1)C(=O)[O-])O

Specifications

Melting Point (decomposition)
Odor Sweet
Quantity 500 g
Solubility Information Soluble in water
Formula Weight 144.05
Grade Technical
Physical Form Powder
Chemical Name or Material Lithium salicylate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 209-011-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only