missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lithium Reagent

Catalog No. TR66056
Change view
Click to view available options
Quantity:
2 x 28 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR66056 2 x 28 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR66056 Supplier Thermo Scientific™ Supplier No. JTR66056

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Assure proper drug dosage is administered in the treatment of illnesses such as manic depression.

Assure proper drug dosage is administered in the treatment of illnesses such as manic depression with Thermo Scientific™ Lithium Reagent.

  • Reagent is a liquid stable assay, offering 12 months stability.
  • Applications are available for most automated chemistry analyzers.

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Description Infinity Lithium
CE Marker Yes
Detectable Analytes Lithium
Sample Type Plasma, Serum
Type Reagent
Linearity 0.04 to 3mmol/L
Methods Compatibility Endpoint
Quantity 2 x 28 mL
Stability Stable Liquid
Form Liquid
Show More Show Less