missing translation for 'onlineSavingsMsg'
Learn More

Lithium Pyruvate Monohydrate 95.0+%, TCI America™

Catalog No. p065925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P065925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P065925G Supplier TCI America Supplier No. P065925G

Chemical Identifiers

CAS 2922-61-4
Molecular Formula C3H3LiO3
Molecular Weight (g/mol) 93.994
MDL Number MFCD00036136
InChI Key OFJHGWPRBMPXCX-UHFFFAOYSA-M
Synonym lithium pyruvate, lithium 2-oxopropanoate, pyruvic acid lithium salt, propanoic acid, 2-oxo-, lithium salt, lithiumpyruvat, lithotab 2-oxopropanoate, propanoic acid, 2-oxo-, lithium salt 1:1, c3h3o3.li, lithium pyruvate h2o, lithium 1+ pyruvate
PubChem CID 23663618
IUPAC Name lithium;2-oxopropanoate
SMILES [Li+].CC(=O)C(=O)[O-]

Specifications

Color White
Quantity 25 g
Formula Weight 93.99
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Pyruvate Monohydrate [Guaranteed for Standard to GOT, GPT]
EINECSNumber (9)-1079
TSCA Yes
Recommended Storage Refrigerator