missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Lithium phenyl-2,4,6-trimethylbenzoylphosphinate

Catalog No. 9008891G
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9008891G 1 g
9008895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9008891G Supplier Sigma Aldrich Supplier No. 9008891G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium phenyl-2,4,6-trimethylbenzoylphosphinate (LAP) is a water soluble, cytocompatible, Type I photoinitiator for use in the polymerization of hydrogels or other polymeric materials. This photoinitator is preferred over Irgacure 2959 for biological applications due to its increased water solubility, increased polymerization rates with 365 nm light, and absorbance at 400 nm allowing for polymerization with visible light. The improved polymerization kinetics enable cell encapsualation at reduced initiator concentration and longer wavelength light, which has been shown to reduce initiator toxicity and increase cell viability.

Specifications

CAS 85073-19-4
Molecular Formula C16H16LiO3P
Linear Formula C16H16LiO3P
Quantity 1 g
Synonym LAP
Molecular Weight (g/mol) 294.21
Percent Purity ≥95%