missing translation for 'onlineSavingsMsg'
Learn More

Lithium Phenyl(2,4,6-trimethylbenzoyl)phosphinate 98.0+%, TCI America™

Catalog No. L02901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L02901G Supplier TCI America Supplier No. L02901G
Only null left

Chemical Identifiers

CAS 85073-19-4
Molecular Formula C16H16LiO3P
Molecular Weight (g/mol) 294.215
InChI Key JUYQFRXNMVWASF-UHFFFAOYSA-M
Synonym Phenyl(2,4,6-trimethylbenzoyl)phosphinic Acid Lithium Salt
PubChem CID 68384915
IUPAC Name lithium;phenyl-(2,4,6-trimethylbenzoyl)phosphinate
SMILES [Li+].CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)[O-])C

Specifications

Color White
Quantity 1 g
Formula Weight 294.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Phenyl(2,4,6-trimethylbenzoyl)phosphinate