missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, Reagent Grade

Catalog No. AA11638A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11638A1 1 kg
AA1163830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11638A1 Supplier Thermo Scientific Chemicals Supplier No. 011638A1

CAS: 13453-78-6 | ClH6LiO7 | 160.431 g/mol

It is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in Baylis-Hillman reaction as a catalyst in the coupling of alpha,beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13453-78-6
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.431
MDL Number MFCD00149765
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
PubChem CID 23708903
IUPAC Name lithium;perchlorate;trihydrate
SMILES [Li+].O.O.O.[O-]Cl(=O)(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 1 kg
Linear Formula LiClO4·3H2O
UN Number UN1481
Merck Index 14,5539
Solubility Information Highly soluble in water,methanol,ethanol,ethyl acetate,diethyl ether,and acetone.
Formula Weight 160.45 (106.40 Anhydrous)
Grade Reagent
Odor Odorless
Density 1.84 g/mL
Chemical Name or Material Lithium perchlorate trihydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 232-237-2
RTECSNumber SC8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only