missing translation for 'onlineSavingsMsg'
Learn More

Lithium Nitrate, Reagent Grade, LabChem™

Catalog No. LC162501
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC162501 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC162501 Supplier LabChem Supplier No. LC162501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent grade lithium nitrate for general laboratory use.

  • Grade: Reagent
  • Physical form: Granules
  • Color: Colorless
  • Molecular formula: LiNO3
  • Formula weight: 68.96
  • Melting point: 264°C
  • Density: 2,380 kg/m3
  • Solubility: Soluble in water
  • Storage: Room temperature

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium(1+) nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Melting Point 264°C
Color Colorless
CAS Max % 100
Physical Form Granules
Quantity 500 g
Linear Formula LiNO3
UN Number UN2722
Solubility Information Soluble in water
InChI Formula InChI=1S/Li.NO3/c;1-2(3)4/q+1;-1
Formula Weight 68.96
Percent Purity ≥98%
Grade Reagent
Identification Passes Test
Packaging Poly Bottle
Product Line LabChem
Appearance Colorless Granules
Specific Gravity 2380kg/m3
Decomposition Information Lithium oxide; Nitrogen oxides
Density 2,380kg/m3
Chemical Name or Material Lithium Nitrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin: Take off contaminated clothing.
Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 232-218-9
Recommended Storage Room Temperature