missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-4407579
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1074209 10 g
AA1074218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1074209 Supplier Thermo Scientific Chemicals Supplier No. 01074209

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol

Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water(2340g/L). Also soluble in ethanol,methanol,pyridine,ammonia and acetone.
IUPAC Name lithium;nitrate
Formula Weight 68.95 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Lithium nitrate hydrate
Hazard Category H272-H302-H319
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 232-218-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only