missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate, anhydrous, 99%

Catalog No. p-4407580
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1340514 25 g
AA1340530 250 g
AA13405A1 1 kg
AA13405A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1340514 Supplier Thermo Scientific Chemicals Supplier No. 01340514
Only null left

Used in Ceramics, Pyrotechnics, Salt Baths | CAS: 7790-69-4 | LiNO3 | 68.94 g/mol

Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point 600°C (decomposition)
Melting Point 264°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99%
Solubility Information Soluble in water,alcohol,ethanol,methanol,pyridine,ammonia and acetone.
IUPAC Name lithium;nitrate
Formula Weight 68.95
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 2.38 g/cm3 at 20°C
Chemical Name or Material Lithium nitrate, anhydrous
Show More Show Less
Hazard Category H272-H302-H319
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 232-218-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only