missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol
Description
Used as a precursor for chemical vapor deposition of thin films for lithium ion battery electrolyte materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Linear Formula | LiNb(OCH3)6 |
| Quantity | 25 mL |
| UN Number | UN1230 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Fully miscible with water. |
| Formula Weight | 285.92 |
| Percent Purity | ≥99% |
| Physical Form | Liquid |
| Assay | (metals basis) |
| Chemical Name or Material | Lithium niobium methoxide |
Safety and Handling
| Hazard Category | H225-H301+H311+H331-H314-H335-H370 |
| Hazard Statement | GHS H Statement H225-H311-H331-H370-H314 Highly flammable liquid and vapor. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs. Causes severe skin burns and eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only