missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol

Catalog No. AA43175AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43175AC 25 mL
AA43175AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43175AC Supplier Thermo Scientific Chemicals Supplier No. 043175AC

CAS: 21864-21-1 | C18H42LiNbO6

Used as solid-state electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21864-21-1
Molecular Formula C18H42LiNbO6
MDL Number MFCD00210626

Specifications

Odor Alcohol-like
Linear Formula LiNb(OC3H7)6
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 454.38
Percent Purity ≥99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lithium niobium isopropoxide
Show More Show Less
Hazard Category H225-H314-H335-H336
Hazard Statement GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only