Learn More
Lithium metaborate, Puratronic™, 99.997% (metals basis)
Description
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 845°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Linear Formula | LiBO2 |
| Solubility Information | Soluble in ethanol. Partially soluble in cold water. |
| IUPAC Name | lithium;oxido(oxo)borane |
| Formula Weight | 49.75 |
| Percent Purity | 99.997% |
| Grade | Puratronic |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 236-631-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only