missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium metaborate, 99%, for analysis
CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
$281.31 - $281.31
Chemical Identifiers
| CAS | 13453-69-5 |
|---|---|
| Molecular Formula | B4H14Li2O7 |
| Molecular Weight (g/mol) | 183.23 |
| MDL Number | MFCD00011089 |
| InChI Key | DPYKRXYVXDYLEY-UHFFFAOYSA-N |
| Synonym | lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs |
| PubChem CID | 123308 |
| IUPAC Name | lithium;oxido(oxo)borane |
| SMILES | [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC365711000
|
Thermo Scientific Chemicals
365711000 |
100 g | Plastic bottle |
Each for $281.31
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13453-69-5 | |
| 183.23 | |
| DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
| 123308 | |
| [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O |
| B4H14Li2O7 | |
| MFCD00011089 | |
| lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
| lithium;oxido(oxo)borane |
Specifications
| 845°C | |
| 13453-69-5 | |
| 100 g | |
| B4H14Li2O7 | |
| MFCD00011089 | |
| Solubility in water: 25.7g/L (20°C) | |
| [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O | |
| 183.23 | |
| 49.75 | |
| Analytical | |
| Lithium metaborate, 99% |
| White | |
| Powder | |
| 99% | |
| LiBO2 | |
| lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
| DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
| lithium;oxido(oxo)borane | |
| 123308 | |
| 99% | |
| Plastic bottle |
Safety and Handling
EINECSNumber : 236-631-5
TSCA : TSCA
RUO – Research Use Only