missing translation for 'onlineSavingsMsg'
Learn More

Lithium L-lactate, 97%

Catalog No. AAA1181818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1181818 50 g
AAA1181830 250 g
AAA118180B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1181818 Supplier Thermo Scientific Chemicals Supplier No. A1181818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27848-80-2 | C3H5LiO3 | 96.01 g/mol

Useful chiral synthon, building block for depsipeptides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27848-80-2
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-DKWTVANSSA-M
Synonym lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li
PubChem CID 23687877
IUPAC Name lithium;(2S)-2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Melting Point 253°C to 255°C
Quantity 50 g
Beilstein 3568332
Sensitivity Hygroscopic
Optical Rotation −14° (c=1 in water)
Formula Weight 96.01
Percent Purity 97%
Chemical Name or Material Lithium L-lactate
EINECSNumber 248-692-5
RTECSNumber OJ6311000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only