missing translation for 'onlineSavingsMsg'
Learn More

Lithium isopropoxide, 99.9% (metals basis)

Catalog No. AA4293004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4293004 2 g
AA4293009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4293004 Supplier Thermo Scientific Chemicals Supplier No. 04293004

CAS: 2388-10-5 | C3H7LiO | 66.03 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 2388-10-5
Molecular Formula C3H7LiO
Molecular Weight (g/mol) 66.03
MDL Number MFCD00048286
InChI Key HAUKUGBTJXWQMF-UHFFFAOYSA-N
Synonym lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
PubChem CID 23673350
IUPAC Name lithium;propan-2-olate
SMILES [Li+].CC(C)[O-]

Specifications

Color White to Yellow
Linear Formula (CH3)2CHOLi
Quantity 2 g
UN Number UN3206
Beilstein 3618614
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 66.03
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium isopropoxide
Show More Show Less
Hazard Category H228-H314-H500
Hazard Statement GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
TSCA No
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only