missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Hydroxide Monohydrate, Honeywell Fluka™
Purum,≥98.5%
$28575.40 - $28575.40
Chemical Identifiers
| CAS | 1310-66-3 |
|---|---|
| Molecular Formula | H3LiO2 |
| Molecular Weight (g/mol) | 41.96 |
| MDL Number | MFCD00149772 |
| InChI Key | GLXDVVHUTZTUQK-UHFFFAOYSA-M |
| Synonym | lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water |
| PubChem CID | 168937 |
| SMILES | [Li+].O.[OH-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6000905
|
Honeywell Chemicals
13020100KG |
100 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 1310-66-3 | |
| 41.96 | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| 168937 |
| H3LiO2 | |
| MFCD00149772 | |
| lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
| [Li+].O.[OH-] |
Specifications
| 1310-66-3 | |
| H3LiO2 | |
| MFCD00149772 | |
| lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
| [Li+].O.[OH-] | |
| 41.96 | |
| 41.96g/mol | |
| purum |
| 100 kg | |
| LiOH · H2O | |
| 2680 | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| lithium;hydroxide;hydrate | |
| 168937 | |
| ≥98.5% | |
| Lithium Hydroxide Monohydrate |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314
EINECSNumber : 215-183-4