missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Hydroxide Monohydrate, ACS Reagent Grade, Ricca Chemical
$210.11 - $210.11
Chemical Identifiers
| CAS | 1310-66-3 |
|---|---|
| Molecular Formula | H3LiO2 |
| Molecular Weight (g/mol) | 41.96 |
| MDL Number | MFCD00149772 |
| InChI Key | GLXDVVHUTZTUQK-UHFFFAOYSA-M |
| IUPAC Name | lithium(1+) hydrate hydroxide |
| SMILES | [Li+].O.[OH-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RDCL0200250
|
Ricca Chemical Company
RDCL0200250A6 |
250 g |
Each for $210.11
|
|
|||||
Chemical Identifiers
| 1310-66-3 | |
| 41.96 | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| [Li+].O.[OH-] |
| H3LiO2 | |
| MFCD00149772 | |
| lithium(1+) hydrate hydroxide |
Specifications
| 1310-66-3 | |
| Fine Crystals or Crystalline Powder | |
| H3LiO2 | |
| GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
| lithium(1+) hydrate hydroxide | |
| min 98% | |
| Natural Poly Bottle |
| 100 | |
| 250 g | |
| MFCD00149772 | |
| [Li+].O.[OH-] | |
| 41.96 | |
| ACS | |
| Lithium Hydroxide Monohydrate |
Safety and Handling
ShelfLife : 84 months