missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCL0200250
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCL0200250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCL0200250 Supplier Ricca Chemical Company Supplier No. RDCL0200250A6
Only null left

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
IUPAC Name lithium(1+) hydrate hydroxide
SMILES [Li+].O.[OH-]

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 250 g
Percent Purity min 98%
Grade ACS
Packaging Natural Poly Bottle
Chemical Name or Material Lithium Hydroxide Monohydrate
ShelfLife 84 months