missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, ACS, 98% min

Catalog No. AA43171A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43171A3 2 kg
AA4317122 100 g
AA4317136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43171A3 Supplier Thermo Scientific Chemicals Supplier No. 043171A3

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium chloride is useful for the production of lithium metal and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 98% min.
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.96 (23.95 Anhydrous)
Grade ACS
Sensitivity Air sensitive; Hygroscopic
Appearance White crystalline powder
Density 1.51
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 215-183-4
RTECSNumber OJ6307080
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only