missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, 99.995% (metals basis)

Catalog No. AA4447336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4447336 500 g
AA4447314 25 g
AA4447322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4447336 Supplier Thermo Scientific Chemicals Supplier No. 04447336

Lithium hydroxide monohydrate, H3LiO2, CAS Number-1310-66-3, unii-g51xlp968g, lithium hydroxide monohydrate, lioh water, lithiumhydrate, hydroxyde de lithium, lithium hydroxido, lithium hydroxide, monohydrate, lithium hydroxide hydrate, lioh.hydrate, lioh-hydrate, 100g, 99.995% (Metals basis), White | CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 41.96 (23.95 Anhydrous)
Density 1.51
Chemical Name or Material Lithium hydroxide monohydrate
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 215-183-4
RTECSNumber OJ6307080
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only