missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Anhydrous 98.0+%, TCI America™

Numéro de catalogue. L0225500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
L0225500G 500 g
L022525G 25 g
L0225100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. L0225500G Fournisseur TCI America Code fournisseur L0225500G
Il en reste null

Identifiants chimiques

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
IUPAC Name lithium(1+) hydroxide
SMILES [Li+].[OH-]

Spécifications

Melting Point 471°C
Color White
Physical Form Powder
Quantity 500 g
UN Number 2680
Formula Weight 23.95
Percent Purity ≥98.0% (T)
Chemical Name or Material Lithium Hydroxide Anhydrous
EINECSNumber (1)-0712
RTECSNumber OJ6307070
TSCA Yes