missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide, 98%+, MilliporeSigma™

Catalog No. p-4939052
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1056910100 100 g Plastic bottle
M1056911000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1056910100 Supplier Technidata Supplier No. 1.05691.0100
Only null left

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
IUPAC Name lithium(1+) hydroxide
SMILES [Li+].[OH-]

Specifications

Physical Form Solid
Quantity 100 g
Assay Percent Range 98%+
UN Number UN 2680
Formula Weight 23.95
Odor Odorless
Packaging Plastic bottle
Appearance Colorless solid
Chemical Name or Material Lithium hydroxide
Hazard Category H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 215-183-4