missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium hexafluoroarsenate(V), 99%
Description
Lithium hexafluoroarsenate(V) is a potential battery electrolyte material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN1557 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | λ⁵-arsenic(5+) lithium(1+) hexafluoride |
| Formula Weight | 195.85 |
| Percent Purity | 99% |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Lithium hexafluoroarsenate(V) |
Safety and Handling
| Hazard Category | H301+H331-H350 |
| Hazard Statement | GHS H Statement H301-H331 Toxic if swallowed. Toxic if inhaled. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S. |
| EINECSNumber | 249-963-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only