missing translation for 'onlineSavingsMsg'
Learn More

Lithium Formate Monohydrate 98.0+%, TCI America™

Catalog No. F0171500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0171500G 500 g
F017125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0171500G Supplier TCI America Supplier No. F0171500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6108-23-2
Molecular Formula CHLiO2
Molecular Weight (g/mol) 51.96
MDL Number MFCD00050823
InChI Key XKPJKVVZOOEMPK-UHFFFAOYSA-M
Synonym lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
PubChem CID 23702287
IUPAC Name lithium(1+) formate
SMILES [Li+].[O-]C=O

Specifications

Color White
Quantity 500 g
Formula Weight 51.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Formate Monohydrate
EINECSNumber (2)-0676
TSCA Yes