missing translation for 'onlineSavingsMsg'
Learn More

Lithium formate hydrate, 98%

Catalog No. AAA1743218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1743218 50 g
AAA1743230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1743218 Supplier Thermo Scientific Chemicals Supplier No. A1743218

CAS: 6108-23-2 | CHLiO2 | 51.96 g/mol

It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6108-23-2
Molecular Formula CHLiO2
Molecular Weight (g/mol) 51.96
MDL Number MFCD00050823
InChI Key XKPJKVVZOOEMPK-UHFFFAOYSA-M
Synonym lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
PubChem CID 23702287
IUPAC Name lithium;formate;hydrate
SMILES [Li+].[O-]C=O

Specifications

Melting Point 94°C (decomposition)
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 51.96 (Anhydrous)
Percent Purity 98%
Odor Mild
Chemical Name or Material Lithium formate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-133-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only