missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium fluoride, 99.98%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1681.0°C |
| Melting Point | 845.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.98% |
| Linear Formula | LiF |
| Fieser | 01,933 |
| Merck Index | 15, 5587 |
| Solubility Information | Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Toxic if swallowed. Causes skin irritation. May cause respiratory irritation. Contact with acids liberates very toxic gas. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. If eye irritation persist |
| EINECSNumber | 232-152- |
RUO – Research Use Only