missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.85% (metals basis)

Catalog No. AA3635909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3635909 10 g
AA3635918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3635909 Supplier Thermo Scientific Chemicals Supplier No. 03635909

CAS: 7789-24-4 | FLi | 25.94 g/mol

Used in the preparation of hypersensitive thermoluminescent materials, dosimeters and in the room-temperature oxidation of noble metals using fluorine in anhydrous HF. It is mainly used as a component of molten salts. Lithium fluoride forms the basic constituent of the preferred fluoride salt mixture used in liquid-fluoride nuclear reactors. Lithium fluoride is widely used in PLED and OLED as a coupling layer to enhance electron injection. LiF crystals are transparent to short wavelength ultraviolet radiation, more so than any other material. LiF is therefore used in specialized UV optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Lump/Powder
Quantity 10 g
Linear Formula LiF
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in 0.29g/100 mL (20°C) and hydrogen fluoride. Insoluble in alcohol.
Formula Weight 25.94
Percent Purity 99.85%
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Assay (metals basis)
Chemical Name or Material Lithium fluoride
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 232-152-0
RTECSNumber OJ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only