missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 98.5%

Catalog No. p-4407565
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1340618 50 g
AA1340630 250 g
AA13406A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1340618 Supplier Thermo Scientific Chemicals Supplier No. 01340618
Only null left

For Use in Vitreous Enamels and Glazes, As Flux in Soldering and Welding Aluminum, Infrared Spectrophotometers | CAS: 7789-24-4 | FLi | 25.94 g/mol

Polyethylene powder is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for manufacturing various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 98.5%
Solubility Information Soluble in water and in acids. Soluble in hydrofluoric acid. Insoluble in alcohol.
IUPAC Name lithium(1+) fluoride
Formula Weight 25.94
Odor Odorless
Refractive Index 1.3915
Appearance White powder
Density 2.635
Chemical Name or Material Lithium fluoride, Volatizes at 1100°-1200°
Show More Show Less
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 232-152-0
RTECSNumber OJ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only