Learn More
Lithium fluoride, 97%, for analysis
CAS: 7789-24-4 | FLi | 25.94 g/mol
Supplier: Thermo Scientific Chemicals 218270025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1681.0°C | |
| 845.0°C | |
| White | |
| 2.5 kg | |
| FLi | |
| MFCD00011090 | |
| 15,5587 | |
| Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol | |
| [Li+].[F-] | |
| 25.94 | |
| 25.93 | |
| Analytical | |
| Plastic bottle | |
| 2.6300g/mL |
| Lithium fluoride | |
| 7789-24-4 | |
| Powder | |
| 96% min | |
| LiF | |
| 01,933 | |
| lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif | |
| PQXKHYXIUOZZFA-UHFFFAOYSA-M | |
| lithium;fluoride | |
| 224478 | |
| 97% | |
| 0.4% (200°C) (typical) | |
| 2.63 |
Chemical Identifiers
| 7789-24-4 | |
| 25.94 | |
| PQXKHYXIUOZZFA-UHFFFAOYSA-M | |
| 224478 |
| FLi | |
| MFCD00011090 | |
| lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif | |
| [Li+].[F-] |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
GHS Signal Word: Danger
EINECSNumber : 232-152-
RUO – Research Use Only