missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium dodecylsulfate, 98%

Catalog No. AAA1600522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1600522 100 g
AAA1600506 5 g
AAA1600514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1600522 Supplier Thermo Scientific Chemicals Supplier No. A1600522
Only null left

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
PubChem CID 2735071
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 100 g
UN Number UN1325
IUPAC Name lithium;dodecyl sulfate
Formula Weight 272.33
Percent Purity 98%
Odor Odorless
Chemical Name or Material Lithium dodecylsulfate
Hazard Category H228-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H318-H228-H335-H315
Causes serious eye damage.
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 218-058-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only