missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Dodecylsulfate, ≥98%
CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol
$189.67 - $485.71
Chemical Identifiers
| CAS | 2044-56-6 |
|---|---|
| Molecular Formula | C12H25LiO4S |
| Molecular Weight (g/mol) | 272.33 |
| MDL Number | MFCD00007467 |
| InChI Key | YFVGRULMIQXYNE-UHFFFAOYSA-M |
| Synonym | lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure |
| PubChem CID | 2735071 |
| SMILES | [Li+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ3281614
|
Thermo Fisher Scientific
J3281614 |
25 g | Plastic bottle |
Each for $189.67
|
|
||||
|
AAJ3281622
|
Thermo Fisher Scientific
J3281622 |
100 g | Plastic bottle |
Each for $485.71
|
|
||||
Chemical Identifiers
| 2044-56-6 | |
| 272.33 | |
| YFVGRULMIQXYNE-UHFFFAOYSA-M | |
| 2735071 |
| C12H25LiO4S | |
| MFCD00007467 | |
| lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure | |
| [Li+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
| 2044-56-6 | |
| 25 g | |
| CH3(CH2)11OSO3Li | |
| UN1325 | |
| YFVGRULMIQXYNE-UHFFFAOYSA-M | |
| lithium;dodecyl sulfate | |
| 2735071 | |
| ≥98% | |
| Lithium Dodecylsulfate |
| Powder | |
| C12H25LiO4S | |
| MFCD00007467 | |
| lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure | |
| [Li+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 272.33 | |
| 272.33 | |
| Plastic bottle |
Safety and Handling
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 4.1
EINECSNumber : 218-058-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only