missing translation for 'onlineSavingsMsg'
Learn More

Lithium Dodecylsulfate, ≥98%

Catalog No. AAJ3281622
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ3281622 100 g Plastic bottle
AAJ3281614 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ3281622 Supplier Thermo Fisher Scientific Supplier No. J3281622
Only null left

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
PubChem CID 2735071
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 100 g
Linear Formula CH3(CH2)11OSO3Li
UN Number UN1325
IUPAC Name lithium(1+) dodecyl sulfate
Formula Weight 272.33
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Lithium Dodecylsulfate
Hazard Category H228-H302+H332-H315-H319-H335
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 218-058-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only