missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Dodecyl Sulfate 98.0+%, TCI America™
$89.48 - $315.03
Chemical Identifiers
| CAS | 2044-56-6 |
|---|---|
| Molecular Formula | C12H25LiO4S |
| Molecular Weight (g/mol) | 272.33 |
| MDL Number | MFCD00007467 |
| InChI Key | YFVGRULMIQXYNE-UHFFFAOYSA-M |
| Synonym | lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure |
| PubChem CID | 2735071 |
| IUPAC Name | lithium(1+) dodecyl sulfate |
| SMILES | [Li+].CCCCCCCCCCCCOS([O-])(=O)=O |
Chemical Identifiers
| 2044-56-6 | |
| 272.33 | |
| YFVGRULMIQXYNE-UHFFFAOYSA-M | |
| 2735071 | |
| [Li+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25LiO4S | |
| MFCD00007467 | |
| lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure | |
| lithium(1+) dodecyl sulfate |
Specifications
| 2044-56-6 | |
| C12H25LiO4S | |
| 5 g | |
| YFVGRULMIQXYNE-UHFFFAOYSA-M | |
| lithium(1+) dodecyl sulfate | |
| 2735071 | |
| ≥98.0% (T) | |
| Lithium Dodecyl Sulfate [for Biochemical Research] |
| White | |
| MFCD00007467 | |
| lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure | |
| [Li+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 272.33 | |
| 272.33 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3045&(8)-0606
TSCA : Yes