missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Dodecyl Sulfate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 272.33 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lithium Dodecyl Sulfate [for Biochemical Research] |
Safety and Handling
| EINECSNumber | (2)-3045&(8)-0606 |
| TSCA | Yes |