missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Lithium dodecyl sulfate, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | (5% aq. soln.) (1 cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm |
| Color | White |
| Physical Form | Shiny Crystalline Flakes or Powder |
| Quantity | 25 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Assay Percent Range | 99+% |
| Linear Formula | CH3(CH2)11OSO3Li |
| Beilstein | 01, IV, 1847 |
| Solubility Information | Solubility in water: soluble. |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: May cause respiratory irritation. Harmful if swallowed. Causes skin irritation. Harmful if inhaled. Causes serious eye irritation. Flammable solid. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only