missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium dodecyl sulfate, 99+%

Catalog No. AC413300250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413300250 25 g Glass bottle
AC413301000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413300250 Supplier Thermo Scientific Chemicals Supplier No. 413300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym Dodecyl lithium sulfate, Lithium lauryl sulfate
PubChem CID 2735071
IUPAC Name lithium(1+) dodecyl sulfate
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance (5% aq. soln.) (1 cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
Color White
Physical Form Shiny Crystalline Flakes or Powder
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99+%
Linear Formula CH3(CH2)11OSO3Li
Beilstein 01, IV, 1847
Solubility Information Solubility in water: soluble.
Formula Weight 272.33
Percent Purity ≥99%
Chemical Name or Material Lithium dodecyl sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only