Learn More
Lithium cobalt(III) oxide, 99.5% (metals basis)
CAS: 12190-79-3 | CoLiO2 | 97.871 g/mol
$201.25 - $201.25
Chemical Identifiers
| CAS | 12190-79-3 |
|---|---|
| Molecular Formula | CoLiO2 |
| Molecular Weight (g/mol) | 97.871 |
| MDL Number | MFCD00049786 |
| InChI Key | BFZPBUKRYWOWDV-UHFFFAOYSA-N |
| Synonym | lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate |
| PubChem CID | 23670860 |
| IUPAC Name | lithium;oxido(oxo)cobalt |
| SMILES | [Li+].[O-][Co]=O |
Description
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehilingâs solution and Nylanderâs test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 12190-79-3 | |
| 97.871 | |
| BFZPBUKRYWOWDV-UHFFFAOYSA-N | |
| 23670860 | |
| [Li+].[O-][Co]=O |
| CoLiO2 | |
| MFCD00049786 | |
| lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate | |
| lithium;oxido(oxo)cobalt |
Specifications
| Blue | |
| Powder | |
| 99.5% (Metal basis) | |
| MFCD00049786 | |
| Insoluble in water. | |
| [Li+].[O-][Co]=O | |
| 97.871 | |
| 97.88 | |
| Hygroscopic | |
| Lithium cobalt(III) oxide |
| 12190-79-3 | |
| 25 g | |
| CoLiO2 | |
| lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate | |
| BFZPBUKRYWOWDV-UHFFFAOYSA-N | |
| lithium;oxido(oxo)cobalt | |
| 23670860 | |
| Odorless | |
| Dark blue powder |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350-H361f
EINECSNumber : 235-362-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only