Learn More
Lithium chloride-potassium chloride
CAS: 65567-96-6 | Cl2KLi | 116.938 g/mol
Supplier: Thermo Scientific Chemicals 01820630
Description
Lithium chloride-potassium chloride is used in separators for mixed transition-metal oxide cathodes in thermal batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lithium chloride-potassium chloride | |
| White | |
| Crystals | |
| Cl2KLi | |
| lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis | |
| HCQWRNRRURULEY-UHFFFAOYSA-L | |
| lithium;potassium;dichloride | |
| 16213218 | |
| Hygroscopic |
| 357°C | |
| 65567-96-6 | |
| 250 g | |
| MFCD00144623 | |
| Soluble in water. | |
| [Li+].[Cl-].[Cl-].[K+] | |
| 116.938 | |
| Odorless | |
| 2.02 |
Chemical Identifiers
| 65567-96-6 | |
| 116.938 | |
| HCQWRNRRURULEY-UHFFFAOYSA-L | |
| 16213218 | |
| [Li+].[Cl-].[Cl-].[K+] |
| Cl2KLi | |
| MFCD00144623 | |
| lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis | |
| lithium;potassium;dichloride |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only