missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate 98.0+%, TCI America™
$42.87 - $227.41
Chemical Identifiers
| CAS | 554-13-2 |
|---|---|
| Molecular Formula | CLi2O3 |
| Molecular Weight (g/mol) | 73.89 |
| MDL Number | MFCD00011084 |
| InChI Key | XGZVUEUWXADBQD-UHFFFAOYSA-L |
| Synonym | lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine |
| PubChem CID | 11125 |
| ChEBI | CHEBI:6504 |
| IUPAC Name | dilithium(1+) carbonate |
| SMILES | [Li+].[Li+].[O-]C([O-])=O |
Chemical Identifiers
| 554-13-2 | |
| 73.89 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| 11125 | |
| dilithium(1+) carbonate |
| CLi2O3 | |
| MFCD00011084 | |
| lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
| CHEBI:6504 | |
| [Li+].[Li+].[O-]C([O-])=O |
Specifications
| 723°C | |
| 554-13-2 | |
| 25 g | |
| MFCD00011084 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| dilithium(1+) carbonate | |
| 11125 | |
| 73.89 | |
| Lithium Carbonate |
| White | |
| Powder | |
| CLi2O3 | |
| lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
| [Li+].[Li+].[O-]C([O-])=O | |
| 73.89 | |
| CHEBI:6504 | |
| ≥98.0% (T) |
Safety and Handling
RTECSNumber : OJ5800000
TSCA : Yes