missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, ACS, 99.0% min

Catalog No. p-4407561
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3622536 500 g
AA36225A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3622536 Supplier Thermo Scientific Chemicals Supplier No. 03622536
Only null left

Maximum level of impurities: Insoluble in dilute hydrochloric acid 0.01%, Cl 0.005%, NO3 5ppm, Sulfur compounds (as SO4) 0.2%, NH4 5ppm, Heavy Metals (as Pb) 0.002%, Fe 0.002%, Ca 0.01%, K 0.01%, Na 0.1% | CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1310°C (decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water and acid. Insoluble in acetone,ammonia and alcohol.
IUPAC Name dilithium;carbonate
Formula Weight 73.9
Grade ACS
Odor Odorless
Appearance White crystalline powder
Density 2.11 g/cm3 at 20°C
Chemical Name or Material Lithium carbonate, For analysis ACS
Show More Show Less
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 209-062-5
RTECSNumber OJ5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only