missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, 99.999%, (trace metal basis), extra pure

Catalog No. p-3756657
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC193360100 10 g Glass bottle
AC193361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC193360100 Supplier Thermo Scientific Chemicals Supplier No. 193360100
Only null left

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 720.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula Li2CO3
Fieser 01,606; 03,183; 04,192; 05,396
Merck Index 15, 5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
IUPAC Name dilithium(1+) carbonate
Formula Weight 73.88
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Density 2.11g/mL
Chemical Name or Material Lithium carbonate, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 209-062-5

RUO – Research Use Only