missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide 99.0+%, TCI America™

Catalog No. p-7131869
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L021025G 25 g
L0210100G 100 g
L0210500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L021025G Supplier TCI America Supplier No. L021025G
Only null left

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Melting Point 549°C
Color White
Physical Form Powder
Quantity 25 g
Formula Weight 86.84
Percent Purity ≥99.0% (T)
Chemical Name or Material Lithium Bromide
EINECSNumber (1)-0110
RTECSNumber OJ5755000
TSCA Yes