missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Bromide 99.0+%, TCI America™
Description
- Purity (Argentometric Titration): ≥99.0%
- Physical State: Solid (white powder to crystal)
- Melting Point: 549°C
- Molecular Weight: 86.84 g/mol
- Solubility: Soluble in acetone; very soluble in water and alcohol
- Storage Temperature: Room temperature (recommended in a cool and dark place, <15°C)
Specifications
Specifications
| Melting Point | 549°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Linear Formula | LiBr |
| Solubility Information | Soluble in Acetone, water, Alcohol |
| InChI Formula | InChI=1S/BrH.Li/h1H;/q;+1/p-1 |
| Formula Weight | 86.84 |
| Percent Purity | ≥99.0% (T) |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| EINECSNumber | (1)-0110 |
| RTECSNumber | OJ5755000 |
| TSCA | Yes |