missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Bromide 99.0+%, TCI America™
Description
Lithium bromide is a white, odorless powder intended for laboratory research. It is suitable for applications such as sodium- and ammonia-free Birch reduction by electroreduction.
- Purity: ≥99.0% (T)
- Chemical Formula: LiBr
- Molecular Weight: 86.84
- Melting Point: 549°C
- Solubility: Very soluble in water; soluble in alcohols and acetone
- Physical State: Powder
Specifications
Specifications
| Melting Point | 549°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.00% |
| Linear Formula | LiBr |
| UN Number | NONH for all modes of transport |
| Solubility Information | Soluble in water and alcohols |
| InChI Formula | InChI=1S/BrH.Li/h1H;/q;+1/p-1 |
| Formula Weight | 86.84 |
| Show More |
Safety and Handling
| EINECSNumber | (1)-0110 |
| RTECSNumber | OJ5755000 |
| TSCA | Yes |