Learn More
Lithium bromide, anhydrous, 99% min
For Pharmaceuticals, Air-Conditioning, Humectant | CAS: 7550-35-8 | BrLi | 86.844 g/mol
Supplier: Thermo Scientific Chemicals 01340822
Description
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange mediumLithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1265°C | |
| 550°C | |
| White | |
| 100 g | |
| BrLi | |
| lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| lithium;bromide | |
| 82050 | |
| 86.85 | |
| 3.464 g/cm3 at 20°C |
| Lithium bromide, anhydrous | |
| 7550-35-8 | |
| Crystals | |
| 99% min. | |
| MFCD00011077 | |
| Very soluble in water and alcohol. Soluble in methanol,acetone,and glycol. Slightly soluble in pyridine. | |
| [Li+].[Br-] | |
| 86.844 | |
| CHEBI:63042 | |
| Hygroscopic |
Chemical Identifiers
| 7550-35-8 | |
| 86.844 | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| 82050 | |
| lithium;bromide |
| BrLi | |
| MFCD00011077 | |
| lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
| CHEBI:63042 | |
| [Li+].[Br-] |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
H302-H315-H317-H319
EINECSNumber : 231-439-8
RTECSNumber : OJ5755000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only