missing translation for 'onlineSavingsMsg'
Learn More

Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™

Catalog No. p-4112117
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428881000 100 mL AcroSeal™ Glass Bottle
AC428888000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428881000 Supplier Thermo Scientific Chemicals Supplier No. 428881000
Only null left

CAS: 16949-15-8 | BH4Li | 21.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Specifications

Chemical Name or Material Lithium Borohydride
CAS 16949-15-8, 109-99-9
Quantity 100 mL
Molecular Formula BH4Li
Linear Formula LiBH4
MDL Number MFCD00011088
Flash Point −21°C
Merck Index 15,5581
Synonym borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1
Solubility Information Solubility in water: decomposes
InChI Key UUKMSDRCXNLYOO-UHFFFAOYSA-N
SMILES [Li+].[BH4-]
IUPAC Name lithium;boron(1-)
Molecular Weight (g/mol) 21.78
PubChem CID 20722760
Formula Weight 21.77
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.89
Density 0.8900g/mL
Show More Show Less

RUO – Research Use Only